Learn More
Cayman Chemical Lanifibranor, Purity: ≥98%, CAS: 927961-18-0, 5 mg
SDP
Catalog No. 503015712
Encompass_Preferred
A PPAR agonist (EC50s = 1,537, 866, and 206 nM for human recombinant PPARα, PPARβ, and PPARγ, respectively); increases β-oxidation in Huh-7 and C2C12 cells at 1 and 3 µM, respectively; reduces plasma glucose and triglyceride levels in a db/db mouse model of type 2 diabetes at 10 and 30 mg/kg for five days; increases plasma adiponectin levels and decreases collagen deposition in a carbon tetrachloride-induced mouse model of NASH
Catalog No. 50-301-5712 Supplier Cayman Chemical Supplier No. 255725 MG
A PPAR agonist (EC50s = 1,537, 866, and 206 nM for human recombinant PPARα, PPARβ, and PPARγ, respectively); increases β-oxidation in Huh-7 and C2C12 cells at 1 and 3 µM, respectively; reduces plasma glucose and triglyceride levels in a db/db mouse model of type 2 diabetes at 10 and 30 mg/kg for five days; increases plasma adiponectin levels and decreases collagen deposition in a carbon tetrachloride-induced mouse model of NASH