Learn More
Selleck Chemical LLC Lanifibranor S8770-10MM/1ML
SDP
Catalog No. 5000023864
Encompass_Preferred
Lanifibranor (IVA-337) is a moderately potent and well balanced pan PPAR agonist with EC50 values of 1537 nM 866 nM and 206 nM for hPPAR hPPAR and hPPAR respectively
Catalog No. 50-000-23864 Supplier Selleck Chemical LLC Supplier No. S877010MM/1ML
Lanifibranor (IVA-337) is a moderately potent and well balanced pan PPAR agonist with EC50 values of 1537 nM 866 nM and 206 nM for hPPAR hPPAR and hPPAR respectively