Learn More
Selleck Chemical LLC Lanifibranor IVA-337-5mg
SDP
Catalog No. 502390030
Encompass_Preferred
Lanifibranor (IVA-337) is a moderately potent and well balanced pan PPAR agonist with EC50 values of 1537 nM, 866 nM and 206 nM for hPPARα, hPPARδ and hPPARγ, respectively.
Catalog No. 50-239-0030 Supplier Selleck Chemical LLC Supplier No. S87705MG
Lanifibranor (IVA-337) is a moderately potent and well balanced pan PPAR agonist with EC50 values of 1537 nM, 866 nM and 206 nM for hPPARα, hPPARδ and hPPARγ, respectively.