Learn More
Medchemexpress LLC HY-14818 10mg , Laninamivir CAS:203120-17-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1481810MG
Encompass_Preferred
HY-14818 10mg Laninamivir CAS: 203120-17-6 Laninamivir (R 125489) is a potent influenza neuraminidase (NA) inhibitor with IC50s of 0.90 nM, 1.83 nM and 3.12 nM for avian H12N5 NA (N5), pH1N1 N1 NA (p09N1) and A/RI/5+/1957 H2N2 N2 (p57N2), respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-1479