Learn More
Medchemexpress LLC Lanoconazole | 101530-10-3 | MFCD00865590 | 98.0% | 319.8 g/mol | C14H10ClN3S2 | 5 MG
SDP
Catalog No. 5000221339
Encompass_Preferred
Lanoconazole is an imidazole-class antifungal research compound supplied as a solid reference standard for laboratory and pharmacological studies. It is used in in vitro and in vivo antifungal testing and mechanistic studies that inhibit sterol 14α-demethylase.
  • Imidazole-class antifungal compound
  • High purity suitable for analytical use
  • Provided as a solid for easy handling and storage
  • Available in small quantities ideal for research
  • Well characterized molecular properties for reference
Catalog No. 50-002-21339 Supplier Medchemexpress LLC Supplier No. HY142825MG
Lanoconazole is an imidazole-class antifungal research compound supplied as a solid reference standard for laboratory and pharmacological studies. It is used in in vitro and in vivo antifungal testing and mechanistic studies that inhibit sterol 14α-demethylase.
  • Imidazole-class antifungal compound
  • High purity suitable for analytical use
  • Provided as a solid for easy handling and storage
  • Available in small quantities ideal for research
  • Well characterized molecular properties for reference