Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lanolin, Anhydrous, Thermo Scientific™

Catalog No. AC412995000
Change view
Click to view available options
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
AC412995000 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC412995000 Supplier Thermo Scientific Chemicals Supplier No. 412995000

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

Acidity 1mg KOH/g max.
CAS 8006-54-0
Infrared Spectrum Authentic
Quantity 500g
MDL Number MFCD00081740
Saponification Value 90 to 105mg KOH/g
Merck Index 15, 5409
Synonym dmt-2'o-tbdms-rc ac phosphoramidite, dmt-2'o-tbdms-rc ac phosphoramidite, configured for abi, dmt-2'o-tbdms-rc ac phosphoramidite, configured for mermade, ac-c-ce phosphoramidite, dmt-2'o-tbdms-rc ac amidite, dmt-2'o-tbdms-rc ac pharmadite r, dmt-2'o-tbdms-rc ac phosphoramidite, configured for ??kta r and oligopilot r, dmt-2'o-tbdms-rc ac phosphoramidite, configured for perkinelmer, configured for polygen, dmt-2'o-tbdms-rc ac phosphoramidite, configured for polygen, configured for perkinelmer
Chemical Name or Material Lanolin
EINECSNumber 232-348-6
RTECSNumber OE3201000
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.