Learn More
TARGETMOL CHEMICALS INC Lanopepden mesylate 1MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T716991MG
Encompass_Preferred
Also available in 5 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Lanopepden mesylate is a peptide deformylase inhibitor against Staphylococcus aureus. Purity 99.44%
Catalog No. 50-287-8205