Learn More
Medchemexpress LLC HY-W020033 1mg Medchemexpress, Lanosterol CAS:79-63-0 Purity:>98%
SDP
Catalog No. 501934650
Encompass_Preferred
Medchemexpress, HY-W020033 1mg Lanosterol CAS:79-63-0 Lanosterol is a key triterpenoid intermediate in the biosynthesis of Cholesterol. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4650 Supplier Medchemexpress LLC Supplier No. HYW0200331MG
Medchemexpress, HY-W020033 1mg Lanosterol CAS:79-63-0 Lanosterol is a key triterpenoid intermediate in the biosynthesis of Cholesterol. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.