Learn More
TARGETMOL CHEMICALS INC LANOSTEROL 50MG
SDP
Catalog No. 502579846
Encompass_Preferred
Also available in 1 mg 100 mg and bulk. Please contact Fisher for quotes. Lanosterol (3(beta)-Hydroxy-824-lanostadiene) is a tetracyclic triterpenoid which is the compound from which all steroids are derived. purity: 99%
Catalog No. 50-257-9846 Supplier TARGETMOL CHEMICALS INC Supplier No. T497050MG
Also available in 1 mg 100 mg and bulk. Please contact Fisher for quotes. Lanosterol (3(beta)-Hydroxy-824-lanostadiene) is a tetracyclic triterpenoid which is the compound from which all steroids are derived. purity: 99%