Learn More
Sigma Aldrich Fine Chemicals Biosciences Lanosterol 93 powder5MG
Catalog No. 502902677
Encompass
Lanosterol an amphipathic molecule that is produced bynbsplanosterolnbspsynthase (LSS). It is enriched in the lens.
Catalog No. 50-290-2677 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. L57685MG
Lanosterol an amphipathic molecule that is produced bynbsplanosterolnbspsynthase (LSS). It is enriched in the lens.