Learn More
Medchemexpress LLC Lanosterol-d6 | 28290-39-3 | MFCD22416700 | >99.0% | 432.75 | C30H44D6O | 1 MG
SDP
Catalog No. 5000301474
Encompass_Preferred
Lanosterol-d6 is a deuterium-labeled analogue of lanosterol provided as a high-purity research reagent for use as an isotopic internal standard and tracer in analytical and biochemical studies. It is optimized for mass spectrometry quantitation, metabolic tracing, and pathway analysis.
  • High isotopic enrichment for accurate mass spectrometry quantitation
  • High purity (>99%) to minimize interfering signals
  • Suitable for use as an internal standard in LC-MS/MS assays
  • Useful for metabolic tracing and pathway studies
  • Stable solid form for easy storage and handling
  • Compatible with common organic solvents for sample preparation
Catalog No. 50-003-01474 Supplier Medchemexpress LLC Supplier No. HYW020033S1MG
Lanosterol-d6 is a deuterium-labeled analogue of lanosterol provided as a high-purity research reagent for use as an isotopic internal standard and tracer in analytical and biochemical studies. It is optimized for mass spectrometry quantitation, metabolic tracing, and pathway analysis.
  • High isotopic enrichment for accurate mass spectrometry quantitation
  • High purity (>99%) to minimize interfering signals
  • Suitable for use as an internal standard in LC-MS/MS assays
  • Useful for metabolic tracing and pathway studies
  • Stable solid form for easy storage and handling
  • Compatible with common organic solvents for sample preparation