Learn More
Medchemexpress LLC Lanosterol-d6 | 28290-39-3 | MFCD22416700 | 99.0% | 432.75 | C30H44D6O | 100 UG
SDP
Catalog No. 5000300265
Encompass_Preferred
Lanosterol-d6 is a deuterium-labeled isotopologue of lanosterol supplied for analytical and research use as an internal standard or tracer in metabolic and quantitative assays. It is intended for use in mass spectrometry and NMR workflows where isotopic labeling improves quantitation and compound tracking.
  • Deuterium-labeled internal standard for quantitative NMR, GC-MS, and LC-MS.
  • High purity, suitable for analytical applications.
  • Molecular formula C30H44D6O and molecular weight 432.75 for reference.
  • Solid form; stable as powder at -20°C for extended storage.
  • Useful for metabolic pathway studies and enzymatic assays related to cholesterol synthesis.
Catalog No. 50-003-00265 Supplier Medchemexpress LLC Supplier No. HYW020033S100UG
Lanosterol-d6 is a deuterium-labeled isotopologue of lanosterol supplied for analytical and research use as an internal standard or tracer in metabolic and quantitative assays. It is intended for use in mass spectrometry and NMR workflows where isotopic labeling improves quantitation and compound tracking.
  • Deuterium-labeled internal standard for quantitative NMR, GC-MS, and LC-MS.
  • High purity, suitable for analytical applications.
  • Molecular formula C30H44D6O and molecular weight 432.75 for reference.
  • Solid form; stable as powder at -20°C for extended storage.
  • Useful for metabolic pathway studies and enzymatic assays related to cholesterol synthesis.