Learn More
TARGETMOL CHEMICALS INC Lanperisone HCl 1MG  DFS Item
Catalog No. 502878221
Also available in 5 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Lanperisone HCl (NK 433) is a novel orally available centrally acting muscle relaxant with anticancer activity and inhibits mono- and polysynaptic reflex potentials.Lanperisone HCl acts by inducing non-apoptotic cell death in the cell cycle and translational independence. Purity 98.67%
Catalog No. 50-287-8221 Supplier TARGETMOL CHEMICALS INC Supplier No. T713561MG
Add to Cart
Add to Cart
Also available in 5 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Lanperisone HCl (NK 433) is a novel orally available centrally acting muscle relaxant with anticancer activity and inhibits mono- and polysynaptic reflex potentials.Lanperisone HCl acts by inducing non-apoptotic cell death in the cell cycle and translational independence. Purity 98.67%