Learn More
Medchemexpress LLC Lanperisone hydrochloride | 116287-13-9 | 99.65% | 1 MG
SDP
Catalog No. 5000234576
Encompass_Preferred
Lanperisone hydrochloride is the hydrochloride salt of lanperisone, a small-molecule research compound studied as an oral muscle relaxant. It modulates mono- and polysynaptic reflex potentials and has shown anticancer and neuroprotective effects in preclinical studies. Supplied as a solid for laboratory research.
  • Hydrochloride salt form for improved solubility.
  • Molecular formula: C15H19ClF3NO; molecular weight: 321.77 g·mol⁻¹.
  • Investigated for muscle relaxation, anticancer, and neuroprotective effects.
  • Distributed in small pack sizes suitable for preclinical research.
Catalog No. 50-002-34576 Supplier Medchemexpress LLC Supplier No. HY119325A1MG
Lanperisone hydrochloride is the hydrochloride salt of lanperisone, a small-molecule research compound studied as an oral muscle relaxant. It modulates mono- and polysynaptic reflex potentials and has shown anticancer and neuroprotective effects in preclinical studies. Supplied as a solid for laboratory research.
  • Hydrochloride salt form for improved solubility.
  • Molecular formula: C15H19ClF3NO; molecular weight: 321.77 g·mol⁻¹.
  • Investigated for muscle relaxation, anticancer, and neuroprotective effects.
  • Distributed in small pack sizes suitable for preclinical research.