Learn More
Medchemexpress LLC Lanraplenib (Synonyms: GS-9876), 100mg | CAS No. 1800046-95-0 | Molecular Weight: 443.50 | Formula: C23H25N9O
SDP
Catalog No. NC2873394
Encompass_Preferred
Lanraplenib (GS-9876) is a highly selective and orally active SYK inhibitor (IC50=9.5 nM) in development for the treatment of inflammatory diseases. Lanraplenib (GS-9876) inhibits SYK activity in platelets via the glycoprotein VI (GPVI) receptor without prolonging bleeding time (BT) in monkeys or humans.
Catalog No. NC2873394 Supplier Medchemexpress LLC Supplier No. HY109091100MG
Lanraplenib (GS-9876) is a highly selective and orally active SYK inhibitor (IC50=9.5 nM) in development for the treatment of inflammatory diseases. Lanraplenib (GS-9876) inhibits SYK activity in platelets via the glycoprotein VI (GPVI) receptor without prolonging bleeding time (BT) in monkeys or humans.