Learn More
Bachem Lanreotide Acetate
SDP
Supplier:  Bachem 4067760.0005
Encompass_Preferred
Lanreotide Acetate, H-D-2-Nal-Cys-Tyr-D-Trp-Lys-Val-Cys-Thr-NH2, 108736-35-2, C54H69N11O10S2, Mr 1096.34, 5mg. BIM-23014, DC-13-116, (D-2-Nal5,Cys6·11,Tyr7,D-Trp8,Val10)-Somatostatin (5-12) amide, Somatostatin Tumor Inhibiting Analog. Lanreotide, a cyclic somatostatin agonist shows a high binding affinity for the somatostatin receptor (SSTR) subtype 2 and has a 10-fold less affinity for receptor subtype 5. It is also a potent inhibitor of growth hormone secretion from pituitary cells. Furthermore, the octapeptide exhibits a direct antiproliferative effect in vivo and in vitro on the growth of cells or solid tumors from a human small cell lung carcinoma. CAS Number (lanreotide acetate): 127984-74-1.
Catalog No. 50-260-217