Learn More
Medchemexpress LLC Lanreotide (acetate) | 2378114-72-6 | 99.8% | 1156.38 | 25 MG
SDP
Catalog No. 5000425480
Encompass_Preferred
Lanreotide (acetate) is a somatostatin analogue with antineoplastic activity, primarily used for the research of carcinoid syndrome. It has demonstrated effects on enhancing radiation-induced apoptosis and inhibiting GH-secreting pituitary adenoma cell proliferation and hormone release in vitro. In vivo studies showed tumor growth inhibition in GH3 tumor-bearing mice.
  • Somatostatin analogue with antineoplastic activity
  • Used for carcinoid syndrome research
  • Enhances radiation-induced apoptosis
  • Inhibits GH-secreting pituitary adenoma cell proliferation and hormone release
  • Produces tumor growth inhibition in vivo
Catalog No. 50-004-25480 Supplier Medchemexpress LLC Supplier No. 25MGHYP1959A
Lanreotide (acetate) is a somatostatin analogue with antineoplastic activity, primarily used for the research of carcinoid syndrome. It has demonstrated effects on enhancing radiation-induced apoptosis and inhibiting GH-secreting pituitary adenoma cell proliferation and hormone release in vitro. In vivo studies showed tumor growth inhibition in GH3 tumor-bearing mice.
  • Somatostatin analogue with antineoplastic activity
  • Used for carcinoid syndrome research
  • Enhances radiation-induced apoptosis
  • Inhibits GH-secreting pituitary adenoma cell proliferation and hormone release
  • Produces tumor growth inhibition in vivo