Learn More
Medchemexpress LLC Lansoprazole-d4 | 934294-22-1 | MFCD06658717 | 99.8% | 373.39 | C16H10D4F3N3O2S | 5 MG
SDP
Catalog No. 5000222437
Encompass_Preferred
Lansoprazole-d4 is a deuterium-labeled analogue of lansoprazole intended as a stable-isotope internal standard for analytical and research use. It is designed for quantitative mass spectrometry, pharmacokinetic studies, and method development to support accurate measurement of lansoprazole in biological and chemical matrices. The compound contains four deuterium atoms and is supplied with high purity for analytical workflows.
  • Deuterium-labeled (d4) analogue for use as an internal standard.
  • High purity for reliable analytical results.
  • Suitable for LC-MS/MS and GC-MS quantification.
  • Enables accurate pharmacokinetic and bioanalysis studies.
  • Supplied in small milligram quantities for analytical workflows.
  • Molecular weight 373.39 and CAS 934294-22-1 for unambiguous identification.
Catalog No. 50-002-22437 Supplier Medchemexpress LLC Supplier No. HY13662S5MG
Lansoprazole-d4 is a deuterium-labeled analogue of lansoprazole intended as a stable-isotope internal standard for analytical and research use. It is designed for quantitative mass spectrometry, pharmacokinetic studies, and method development to support accurate measurement of lansoprazole in biological and chemical matrices. The compound contains four deuterium atoms and is supplied with high purity for analytical workflows.
  • Deuterium-labeled (d4) analogue for use as an internal standard.
  • High purity for reliable analytical results.
  • Suitable for LC-MS/MS and GC-MS quantification.
  • Enables accurate pharmacokinetic and bioanalysis studies.
  • Supplied in small milligram quantities for analytical workflows.
  • Molecular weight 373.39 and CAS 934294-22-1 for unambiguous identification.