Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lansoprazole Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111002439 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-100-2439 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-100-2439 Supplier MilliporeSigma Supelco Supplier No. PHR13901G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Lansoprazole is an important proton pump inhibitor and a substituted benzimidazole with antisecretory and antiulcer activities. Its mode of action involves the suppression of gastric acid secretion by specific inhibition of the gastric H+/K+ ATPase enzyme system at the secretory surface of the gastric parietal cells.Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.
TRUSTED_SUSTAINABILITY

Specifications

CAS 103577-45-3
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.
Physical Form Neat
Linear Formula C16H14F3N3O2 S
MDL Number MFCD00866873
Synonym Lansoprazole
Type Certified Reference Material
Quantity 1 g
Formula Weight 369.36
Grade Certified Reference Material
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.