Learn More
Medchemexpress LLC Lansoprazole Sulfide | 103577-40-8 | 99.9% | 353.37 | 1 G
SDP
Catalog No. 5000454181
Encompass_Preferred
Lansoprazole sulfide is an active metabolite of Lansoprazole, formed by cytochrome P450. It demonstrates anti-cancer activity and acts as an anti-tuberculous agent.
  • Possesses anti-cancer activity
  • Acts as an anti-tuberculous agent, with an IC50 of 0.59 μM for M. tb.
Catalog No. 50-004-54181 Supplier Medchemexpress LLC Supplier No. HYW0131861G
Lansoprazole sulfide is an active metabolite of Lansoprazole, formed by cytochrome P450. It demonstrates anti-cancer activity and acts as an anti-tuberculous agent.
  • Possesses anti-cancer activity
  • Acts as an anti-tuberculous agent, with an IC50 of 0.59 μM for M. tb.