Learn More
Medchemexpress LLC Lansoprazole Sulfide | 103577-40-8 | 99.9% | 353.37 | 1 ML
SDP
Catalog No. 5000454417
Encompass_Preferred
Lansoprazole sulfide is an active metabolite of Lansoprazole, generated by cytochrome P450. It exhibits anti-cancer activity and acts as an anti-tuberculous agent with an IC50 of 0.59 μM for M. tb. It is intended for research use only.
  • Active metabolite of Lansoprazole.
  • Possesses anti-cancer activity.
  • Anti-tuberculous agent with an IC50 of 0.59 μM for M. tb.
Catalog No. 50-004-54417 Supplier Medchemexpress LLC Supplier No. HYW01318610MM/1ML
Lansoprazole sulfide is an active metabolite of Lansoprazole, generated by cytochrome P450. It exhibits anti-cancer activity and acts as an anti-tuberculous agent with an IC50 of 0.59 μM for M. tb. It is intended for research use only.
  • Active metabolite of Lansoprazole.
  • Possesses anti-cancer activity.
  • Anti-tuberculous agent with an IC50 of 0.59 μM for M. tb.