Learn More
Medchemexpress LLC Lansoprazole sulfide | 103577-40-8 | 99.9% | 353.37 | 250MG
SDP
Catalog No. 5000366304
Encompass_Preferred
Lansoprazole sulfide is an active metabolite of Lansoprazole, generated by cytochrome P450. This compound exhibits significant anti-cancer activity and serves as an effective anti-tuberculous agent against M. tb.
  • Active metabolite of Lansoprazole.
  • Exhibits anti-cancer activity.
  • Functions as an anti-tuberculous agent with an IC50 of 0.59 μM for M. tb.
Catalog No. 50-003-66304 Supplier Medchemexpress LLC Supplier No. 250MGHYW013186
Lansoprazole sulfide is an active metabolite of Lansoprazole, generated by cytochrome P450. This compound exhibits significant anti-cancer activity and serves as an effective anti-tuberculous agent against M. tb.
  • Active metabolite of Lansoprazole.
  • Exhibits anti-cancer activity.
  • Functions as an anti-tuberculous agent with an IC50 of 0.59 μM for M. tb.