Learn More
Medchemexpress LLC Lansoprazole sulfide | 103577-40-8 | 99.9% | 353.37 | 500 MG
SDP
Catalog No. 5000454405
Encompass_Preferred
Lansoprazole sulfide is an active metabolite of Lansoprazole by cytochrome P450, possessing anti-cancer activity. It functions as an anti-tuberculous agent, exhibiting an IC50 of 0.59 μM for *M. tb*.
  • Active metabolite of Lansoprazole by cytochrome P450.
  • Possesses anti-cancer activity.
  • Anti-tuberculous agent with an IC50 of 0.59 μM for *M. tb*.
Catalog No. 50-004-54405 Supplier Medchemexpress LLC Supplier No. HYW013186500MG
Lansoprazole sulfide is an active metabolite of Lansoprazole by cytochrome P450, possessing anti-cancer activity. It functions as an anti-tuberculous agent, exhibiting an IC50 of 0.59 μM for *M. tb*.
  • Active metabolite of Lansoprazole by cytochrome P450.
  • Possesses anti-cancer activity.
  • Anti-tuberculous agent with an IC50 of 0.59 μM for *M. tb*.