Learn More
Medchemexpress LLC Lansoprazole sulfone | 131926-99-3 | 99.5% | 385.36 | 10 MG
SDP
Catalog No. 5000452690
Encompass_Preferred
Lansoprazole sulfone (AG-1813) is an orally active and selective inhibitor of H+, K+-ATPase. It significantly stimulates gastric acid secretion by inhibiting H+, K+-ATPase. This compound has potential applications in treating various gastrointestinal conditions.
  • Orally active
  • Selective H+, K+-ATPase inhibitor
  • Stimulates gastric acid secretion
  • Potential application in duodenal ulcer
  • Potential application in gastric ulcer
  • Potential application in gastroesophageal reflux disease
  • Potential application in Zollinger-Ellison disease
Catalog No. 50-004-52690 Supplier Medchemexpress LLC Supplier No. HYW00861410MG
Lansoprazole sulfone (AG-1813) is an orally active and selective inhibitor of H+, K+-ATPase. It significantly stimulates gastric acid secretion by inhibiting H+, K+-ATPase. This compound has potential applications in treating various gastrointestinal conditions.
  • Orally active
  • Selective H+, K+-ATPase inhibitor
  • Stimulates gastric acid secretion
  • Potential application in duodenal ulcer
  • Potential application in gastric ulcer
  • Potential application in gastroesophageal reflux disease
  • Potential application in Zollinger-Ellison disease