Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Lansoprazole sulfone | 131926-99-3 | 99.5% | 385.36 | 10 MM * 1 ML
SDP

Catalog No. 5000366585
Encompass_Preferred

Lansoprazole sulfone is an orally active and selective inhibitor of H+, K+-ATPase. It significantly stimulates gastric acid secretion by inhibiting H+, K+-ATPase and has potential applications in duodenal ulcer, gastric ulcer, gastroesophageal reflux disease, and Zollinger-Ellison disease.

  • Orally active and selective inhibitor of H+, K+-ATPase.
  • Stimulates gastric acid secretion by inhibiting H+, K+-ATPase.
  • Potential applications in duodenal ulcer, gastric ulcer, gastroesophageal reflux disease, and Zollinger-Ellison disease.
  • For research use only.
  • Appearance: Solid, white to off-white.
  • Shipping: Room temperature in continental US; may vary elsewhere.
  • Storage: 4°C, sealed storage, away from moisture. In solvent: -80°C for 6 months; -20°C for 1 month (sealed storage, away from moisture).

Catalog No. 50-003-66585 Supplier Medchemexpress LLC Supplier No. HYW00861410MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Lansoprazole sulfone is an orally active and selective inhibitor of H+, K+-ATPase. It significantly stimulates gastric acid secretion by inhibiting H+, K+-ATPase and has potential applications in duodenal ulcer, gastric ulcer, gastroesophageal reflux disease, and Zollinger-Ellison disease.

  • Orally active and selective inhibitor of H+, K+-ATPase.
  • Stimulates gastric acid secretion by inhibiting H+, K+-ATPase.
  • Potential applications in duodenal ulcer, gastric ulcer, gastroesophageal reflux disease, and Zollinger-Ellison disease.
  • For research use only.
  • Appearance: Solid, white to off-white.
  • Shipping: Room temperature in continental US; may vary elsewhere.
  • Storage: 4°C, sealed storage, away from moisture. In solvent: -80°C for 6 months; -20°C for 1 month (sealed storage, away from moisture).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.