Learn More
Medchemexpress LLC Lansoprazole sulfone | 131926-99-3 | 99.5% | 385.36 | 100 MG
SDP
Catalog No. 5000366587
Encompass_Preferred
Lansoprazole sulfone (AG-1813) is an orally active and selective inhibitor of H+, K+-ATPase. It significantly stimulates gastric acid secretion by inhibiting H+, K+-ATPase. It has potential applications in duodenal ulcer, gastric ulcer, gastroesophageal reflux disease, and Zollinger-Ellison disease. It is for research use only and not sold to patients.
  • Orally active and selective inhibitor of H+, K+-ATPase.
  • Significantly stimulates gastric acid secretion by inhibiting H+, K+-ATPase.
  • Potential applications in duodenal ulcer, gastric ulcer, gastroesophageal reflux disease, and Zollinger-Ellison disease.
Catalog No. 50-003-66587 Supplier Medchemexpress LLC Supplier No. HYW008614100MG
Lansoprazole sulfone (AG-1813) is an orally active and selective inhibitor of H+, K+-ATPase. It significantly stimulates gastric acid secretion by inhibiting H+, K+-ATPase. It has potential applications in duodenal ulcer, gastric ulcer, gastroesophageal reflux disease, and Zollinger-Ellison disease. It is for research use only and not sold to patients.
  • Orally active and selective inhibitor of H+, K+-ATPase.
  • Significantly stimulates gastric acid secretion by inhibiting H+, K+-ATPase.
  • Potential applications in duodenal ulcer, gastric ulcer, gastroesophageal reflux disease, and Zollinger-Ellison disease.