Learn More
Medchemexpress LLC Lansoprazole sulfone | 131926-99-3 | 99.5% | 385.36 | 5 MG
SDP
Catalog No. 5000366588
Encompass_Preferred
Lansoprazole sulfone is an orally active and selective inhibitor of H+, K+-ATPase. It can significantly stimulate gastric acid secretion by inhibiting H+, K+-ATPase. This compound has potential applications in duodenal ulcer, gastric ulcer, gastroesophageal reflux disease, and Zolinger Ellison disease.
  • Orally active
  • Selective inhibitor of H+, K+-ATPase
  • Significantly stimulates gastric acid secretion by inhibiting H+, K+-ATPase
  • Potential applications in duodenal ulcer, gastric ulcer, gastroesophageal reflux disease, and Zolinger Ellison disease
Catalog No. 50-003-66588 Supplier Medchemexpress LLC Supplier No. HYW0086145MG
Lansoprazole sulfone is an orally active and selective inhibitor of H+, K+-ATPase. It can significantly stimulate gastric acid secretion by inhibiting H+, K+-ATPase. This compound has potential applications in duodenal ulcer, gastric ulcer, gastroesophageal reflux disease, and Zolinger Ellison disease.
  • Orally active
  • Selective inhibitor of H+, K+-ATPase
  • Significantly stimulates gastric acid secretion by inhibiting H+, K+-ATPase
  • Potential applications in duodenal ulcer, gastric ulcer, gastroesophageal reflux disease, and Zolinger Ellison disease