Learn More
TARGETMOL CHEMICALS INC Lansoprazole sulfone 5MG
SDP
Catalog No. 502877458
Encompass_Preferred
Also available in 1 mL, 1 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Lansoprazole sulfone is a metabolite of the gastric pump inhibitor Lansoprazole. Purity 100%
Catalog No. 50-287-7458 Supplier TARGETMOL CHEMICALS INC Supplier No. T201555MG
Also available in 1 mL, 1 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Lansoprazole sulfone is a metabolite of the gastric pump inhibitor Lansoprazole. Purity 100%