Learn More
Selleck Chemical LLC Lanthanum
SDP
Supplier:  Selleck Chemical LLC S551425MG
Encompass_Preferred
Lanthanum(III) chloride is the inorganic compound used in biochemical research to block the activity of divalent cation channels mainly calcium channels
Catalog No. 50-000-04052