Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Lanthanum boride

Catalog No. 7091905G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
70-919-05G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 70-919-05G Supplier Sigma Aldrich Supplier No. 7091905G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Lanthanum boride

Specifications

Melting Point 2210°C
CAS 12008-21-8
Quantity 5 g
Molecular Formula B6La
Linear Formula LaB6
MDL Number MFCD00151350
Synonym Lanthanum hexaboride; Lanthanum boride
Molecular Weight (g/mol) 203.77
Percent Purity 99.5% trace metals basis
Density 2.61 g/cm3 at 25°C; 4.76 g/cm3 (at 25°C)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.