Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lanthanum boride, 99.5% (REO)

Catalog No. AA4032509
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4032509 10 g
AA4032518 50 g
AA4032530 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA4032509 Supplier Thermo Scientific Chemicals Supplier No. 04032509
Add to Cart

CAS: 12008-21-8 | B6La-2 | 203.765 g/mol

Lanthanum boride is employed in hot cathodes, which enact and includes electron microscopes, microwave tubes, electron lithography, electron beam welding, X-ray tubes, and free electron lasers. It is administered as a size or strain standard in X-ray powder diffraction to calibrate instrumental broadening of diffraction peaks. It exercises as a superconductor. It has a great potential as a thermionic emitter in renewable energy applications. It is widely used in teleran, electronic industry, instrument, medical equipment, metallurgy and environmental protection.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12008-21-8
Molecular Formula B6La-2
Molecular Weight (g/mol) 203.765
MDL Number MFCD00151350
InChI Key IBDHMAYMLSYGAK-UHFFFAOYSA-N
Synonym lanthanum boride, lanthanum hexaboride, lanthanum boride reo, lanthanum hexaboride lab6 grade a, lanthanum hexaboride lab6 grade b, lanthanum hexaboride lab6 grade c, lanthanum hexaboride, powder, 10 mum, lanthanum boride, powder,-325 mesh trace metals basis, pentacyclo 3.1.0.0 1 , 3 .0 2 ,?.0?,? hexaborane-1,4-diuide lanthanum, line position and line shape standard for powder diffraction, nist srm 660c, lanthanum hexaboride powder
PubChem CID 71308229
SMILES B12B3[B-]14B5[B-]23B45.[La]

Specifications

Melting Point 2210°C
Physical Form Powder
Quantity 10 g
Solubility Information Insoluble in water,HCl.
Formula Weight 203.78
Percent Purity 99.5%
Odor Odorless
Density 2.61 g/mL
Assay (REO)
Chemical Name or Material Lanthanum boride
EINECSNumber 234-531-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.