Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lanthanum Chloride Saturated Hydrate, 64.5-70.0%, (ACS Reagent Grade), MP Biomedicals

Catalog No. ICN15253490 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15253490 500 g
ICN15253425 25 g
ICN15253480 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN15253490 Supplier MP Biomedicals, Inc Supplier No. 0215253490
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 10025-84-0 | Cl3H14LaO7 | 371.36 g/mol

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10025-84-0
Molecular Formula Cl3H14LaO7
Molecular Weight (g/mol) 371.36
MDL Number MFCD00149756
InChI Key FDFPDGIMPRFRJP-UHFFFAOYSA-K
IUPAC Name lanthanum(3+) heptahydrate trichloride
SMILES O.O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[La+3]

Specifications

Color White
Quantity 500 g
Assay Percent Range 64.5-70.0%
Synonym Lanthanum Trichloride, Lanthanum III Chloride
Formula Weight 371.4
Grade ACS Reagent
Appearance Crystalline Powder
Chemical Name or Material Lanthanum Chloride Saturated Hydrate
Recommended Storage Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.