Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lanthanum Hexaboride 99%, Thermo Scientific™

Catalog No. AC221380250
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC221380250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC221380250 Supplier Thermo Scientific Chemicals Supplier No. 221380250

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12008-21-8
Molecular Formula B6La-2
Molecular Weight (g/mol) 203.765
InChI Key IBDHMAYMLSYGAK-UHFFFAOYSA-N
Synonym lanthanum boride, lanthanum hexaboride, lanthanum boride reo, lanthanum hexaboride lab6 grade a, lanthanum hexaboride lab6 grade b, lanthanum hexaboride lab6 grade c, lanthanum hexaboride, powder, 10 mum, lanthanum boride, powder,-325 mesh trace metals basis, pentacyclo 3.1.0.0 1 , 3 .0 2 ,?.0?,? hexaborane-1,4-diuide lanthanum, line position and line shape standard for powder diffraction, nist srm 660c, lanthanum hexaboride powder
PubChem CID 71308229
SMILES B12B3[B-]14B5[B-]23B45.[La]

Specifications

Color Blue
Physical Form Powder
Quantity 25 g
Linear Formula LaB6
Formula Weight 203.78
Percent Purity ≥98.5%
Density 2.61
Chemical Name or Material Lanthanum hexaboride, 99%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.