Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lanthanum(III) 2-methoxyethoxide, 99.9% (REO), 5% w/v in 2-methoxyethanol

Catalog No. AA42522AC
Change view
Click to view available options
Quantity:
25 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA42522AC 25 mL
AA42522AE 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA42522AC Supplier Thermo Scientific Chemicals Supplier No. 042522AC
Add to Cart

Preparation of lanthanum oxide thin films by the Sol-Gel method | CAS: 115685-56-8

Lanthanum(III) 2-methoxyethoxide is used in the preparation of lanthanum oxide thin films by the Sol-Gel method

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 115685-56-8
MDL Number MFCD00210619
Synonym lanthanum iii 2-methoxyethoxide, lanthanum tris 1-methoxyethan-1-olate, acmc-20akoi, lanthanum 3+ ion tris 1-methoxyethanolate

Specifications

Color Yellow
Density 0.96 g/mL
Boiling Point 124°C
Flash Point 40°C (104°F)
Linear Formula La(OCH2CH2OCH3)3
Quantity 25 mL
UN Number UN1188
Sensitivity Moisture sensitive
Solubility Information Hydrolyzes in water.
Formula Weight 364.17
Percent Purity 99.9%
Concentration or Composition (by Analyte or Components) 5% w/v in 2-methoxyethanol
Physical Form Liquid
Assay (REO)
Chemical Name or Material Lanthanum (III) 2-methoxyethoxide
Show More Show Less
Hazard Category H226-H302+H312+H332-H360FD-H370-H373
Hazard Statement GHS H Statement
H360-H226-H302-H312-H332
May damage fertility or the unborn child.
Flammable liquid and vapor.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Precautionary Statement P201-P202-P210-P233-P240-P241-P242-P243-P260-P264b-P270-P271-P281-P301+P312-P303+P361+P353-P304+P340-P307+P311-P312-P330-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ETHYLENE GLYCOL MONOMETHYL ETHER
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.