Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Lanthanum(III) acetate hydrate

Catalog No. 306339100G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
30-633-9100G 100 g
30-633-9500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 30-633-9100G Supplier Sigma Aldrich Supplier No. 306339100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Used in the synthesis of a novel phenolate-bridged dilanthanum(III) complex which has potential as a model for metalloproteins as well as in basic and applied chemistry.

Specifications

CAS 100587-90-4
Molecular Formula C6H9LaO6 -+ xH2O
Linear Formula La(CH3CO2)3 -+ xH2O
MDL Number MFCD00150118
Quantity 100 g
Molecular Weight (g/mol) 316.04
Percent Purity 99.9%trace rare earth metals basis
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.