Learn More
Strem, An Ascensus Company CAS# 64424-12-0. 25g. Lanthanum(III) acetylacetonate hydrate (99.9%-La) (REO). MFCD00149057
SDP
Catalog No. 5090115825
Encompass
CAS# 64424-12-0. 25g. Lanthanum(III) acetylacetonate hydrate (99.9%-La) (REO). MFCD00149057. Molecular Weight: 436.24. Molecular Formula: La(CH3C(O)CHC(O)CH3)3.XH2O. Color/form: white to off-white pwdr. Strem# 93-5702. http://www.strem.com/catalog/v/93-5702/
Catalog No. 50-901-15825 Supplier Strem, An Ascensus Company Supplier No. 93570225G
CAS# 64424-12-0. 25g. Lanthanum(III) acetylacetonate hydrate (99.9%-La) (REO). MFCD00149057. Molecular Weight: 436.24. Molecular Formula: La(CH3C(O)CHC(O)CH3)3.XH2O. Color/form: white to off-white pwdr. Strem# 93-5702. http://www.strem.com/catalog/v/93-5702/