Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Lanthanum(III) Chloride Bis(Lithium Chloride) Complex Solution

Catalog No. 703559100ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
70-355-9100ML 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Danger
Catalog No. 70-355-9100ML Supplier Sigma Aldrich Supplier No. 703559100ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Selective 1,2-Additions with LaCl3.2LiClMediates Grignard additionsReagent for metal exchange reactionsPromoter for the addition of organometallic compounds to sterically hindered, enolizable or +a,+a-unsaturated ketones or iminesProduct of Albemarle US Inc

Specifications

CAS 71360-06-0
Density 1.017 g/mL at 25°C
Molecular Formula C16H19P
Linear Formula C16H19P
MDL Number MFCD01074629
Quantity 500 mg
Molecular Weight (g/mol) 242.3 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.