Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lanthanum(III) chloride heptahydrate 98%, MilliporeSigma™

Catalog No. M1122190100 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 g
Packaging:
Plastic Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
M1122190100 100 g Plastic Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M1122190100 Supplier MilliporeSigma Supplier No. 1.12219.0100
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

High-purity grade lanthanum(III) chloride heptahydrate for analytical applications.

  • Lanthanum (III) chloride heptahydrate 98%, reagent for analysis, supplied in a 100 g plastic bottle
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10025-84-0 , 7732-18-5
Molecular Formula Cl3H14LaO7
Molecular Weight (g/mol) 371.36
MDL Number MFCD00149756
InChI Key FDFPDGIMPRFRJP-UHFFFAOYSA-K
Synonym Lanthanum trichloride
IUPAC Name lanthanum(3+) heptahydrate trichloride
SMILES O.O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[La+3]

Specifications

Melting Point 91°C
Color White
pH 5
Physical Form Solid
Quantity 100 g
Assay Percent Range 98%
Linear Formula LaCl3 · 7H2O
Solubility Information Soluble (20°C)
Formula Weight 371.37
Identification Passes Test
Packaging Plastic Bottle
Appearance White Solid
Density Bulk Density: 900 kg/m3
Chemical Name or Material For Analysis
Show More Show Less
Recommended Storage Store without restrictions
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.