Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lanthanum(III) chloride heptahydrate, Puriss. p.a., ≥99%, Solstice

Catalog No. 6002390 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
1 Ea.
100 g
6 x 100 g
Packaging:
Ampoule
Poly bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-023-90 100 g Poly bottle
60-045-978 6 x 100 g Poly bottle
60-024-03 1 Ea. Ampoule
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 60-023-90 Supplier Solstice Supplier No. 31820100G
Add to Cart
Add to Cart

for 1L standard solution

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10025-84-0

Specifications

Quantity 100 g
Molecular Formula Cl3H14LaO7
Linear Formula LaCl3 · 7H2O
MDL Number MFCD00149756
UN Number NONH for all modes of transport
InChI Key FDFPDGIMPRFRJP-UHFFFAOYSA-K
SMILES O.O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[La+3]
IUPAC Name lanthanum(3+) heptahydrate trichloride
Molecular Weight (g/mol) 371.36
Formula Weight 371.37g/mol
Percent Purity ≥99%
Grade puriss. p.a.
Packaging Poly bottle
Chemical Name or Material Lanthanum(Iii) Chloride Heptahydrate
Show More Show Less
Hazard Statement P261-P305 + P351 + P338
Precautionary Statement H315-H319-H335
RTECSNumber OE4375000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.