Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lanthanum(III) chloride hydrate, 99.9% (REO)

Catalog No. AA4312036
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4312036 500 g
AA4312022 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4312036 Supplier Thermo Scientific Chemicals Supplier No. 04312036
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 20211-76-1 | Cl3La | 245.26 g/mol

Lanthanum(III) chloride is useful as precursor for synthesis of aqueous sol-gel lanthanum phosphate nano rods,as a filter aid,an effective flocculent and precipitation phosphate. It is also used as a catalyst for the high pressure oxidative chlorination of methane to chloromethane and organic synthesis it acts as Lewis acid catalyst. It is used as a scintillator material and in Gamma Detectors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20211-76-1
Molecular Formula Cl3La
Molecular Weight (g/mol) 245.26
MDL Number MFCD00149753
InChI Key ICAKDTKJOYSXGC-UHFFFAOYSA-K
IUPAC Name lanthanum(3+) trichloride
SMILES [Cl-].[Cl-].[Cl-].[La+3]

Specifications

Melting Point 91°C (decomposition)
pH 5
Physical Form Crystalline
Quantity 500 g
Odor Odorless
Merck Index 14,5363
Solubility Information Soluble in water and ethanol.
Formula Weight 245.27 (Anhydrous)
Percent Purity 99.9%
Assay (REO)
Chemical Name or Material Lanthanum (III) chloride hydrate
Show More Show Less
Hazard Category H290-H317-H318
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P234-P261-P272-P280-P302+P352-P305+P351+P338-P310-P333+P313-P363-P390-P501c
DOTInformation Hazard Class: 8; Packaging Group: III
EINECSNumber 233-237-5
RTECSNumber OE4470000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.