Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lanthanum(III) chloride, ultra dry, 99.9% (REO)

Catalog No. AA3570230
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3570230 250 g
AA3570209 10 g
AA3570218 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA3570230 Supplier Thermo Scientific Chemicals Supplier No. 03570230
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 10099-58-8 | Cl3La | 245.26 g/mol

Lanthanum chloride solutions are used in various water treatment applications. It is also used in biochemical research. Doped with cerium, it is applied as a scintillator material. In organic synthesis, lanthanum trichloride functions as a mild lewis acid. This compound is used in gamma detectors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10099-58-8
Molecular Formula Cl3La
Molecular Weight (g/mol) 245.26
MDL Number MFCD00011068
InChI Key ICAKDTKJOYSXGC-UHFFFAOYSA-K
IUPAC Name lanthanum(3+) trichloride
SMILES [Cl-].[Cl-].[Cl-].[La+3]

Specifications

Boiling Point 1,000°C
Melting Point 860°C
Color White
Physical Form Beads
Quantity 250 g
Odor Odorless
Packaging Ampule
Sensitivity Hygroscopic
Density 3.842 g/mL
UN Number UN3077
Merck Index 14,5363
Solubility Information Soluble in water.
Formula Weight 245.27
Percent Purity 99.9%
Assay (REO)
Chemical Name or Material Lanthanum(III) chloride, Ultra dry
Show More Show Less
Hazard Category H290-H317-H318
Hazard Statement GHS H Statement
H318-H290-H317
Causes serious eye damage.
May be corrosive to metals.
May cause an allergic skin reaction.
Precautionary Statement P234-P261-P272-P280-P302+P352-P305+P351+P338-P310-P333+P313-P363-P390-P501c
DOTInformation Hazard Class: 8; Packaging Group: III
EINECSNumber 233-237-5
RTECSNumber OE4375000
TSCA Yes
storageNote1
  • Ampouled under argon
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.