Learn More
Strem, An Ascensus Company CAS# 10277-43-7. 500g. Lanthanum(III) nitrate hexahydrate (99.99%-La) (REO) PURATREM. MFCD00149751
SDP
Catalog No. 5090115853
Encompass
CAS# 10277-43-7. 500g. Lanthanum(III) nitrate hexahydrate (99.99%-La) (REO) PURATREM. MFCD00149751. Molecular Weight: 324.93 (433.02). Molecular Formula: La(NO3)3∙6H2O. Color/form: white xtl. Strem# 93-5732. http://www.strem.com/catalog/v/93-5732/
Catalog No. 50-901-15853 Supplier Strem, An Ascensus Company Supplier No. 935732500G
CAS# 10277-43-7. 500g. Lanthanum(III) nitrate hexahydrate (99.99%-La) (REO) PURATREM. MFCD00149751. Molecular Weight: 324.93 (433.02). Molecular Formula: La(NO3)3∙6H2O. Color/form: white xtl. Strem# 93-5732. http://www.strem.com/catalog/v/93-5732/