Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Lanthanum(III) nitrate hexahydrate

Catalog No. 3319375G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
33-193-75G 5 g
33-193-7500G 500 g
33-193-7100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 33-193-75G Supplier Sigma Aldrich Supplier No. 3319375G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Lanthanum(III) nitrate hexahydrate

Specifications

Melting Point 65°C to 68°C
CAS 10277-43-7
Quantity 5 g
Molecular Formula LaN3O9 -+ 6H2O
Linear Formula La(NO3)3 -+ 6H2O
MDL Number MFCD00149751
Synonym Nitric acid, lanthanum (III) salt, hexahydrate
Molecular Weight (g/mol) 433.01
Percent Purity 99.99% trace metals basis
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.