Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lanthanum(III) oxalate hydrate, 99.9% (REO)

Catalog No. AA4312222
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4312222 100 g
AA4312236 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4312222 Supplier Thermo Scientific Chemicals Supplier No. 04312222
Add to Cart

CAS: 79079-18-8 | C6La2O12 | 541.87 g/mol

Lanthanum Oxalate is mainly applied in specialty glass, phosphors, water treatment and catalyst. This material use in glass, ceramics, paper, textile, coating, fine and specialty chemicals. In petroleum cracking catalysts.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 79079-18-8
Molecular Formula C6La2O12
Molecular Weight (g/mol) 541.87
MDL Number MFCD00150540
InChI Key OXHNIMPTBAKYRS-UHFFFAOYSA-H
Synonym lanthanum iii oxalate decahydrate, lanthanum oxalate decahydrate, lanthanum iii oxalate, bis 4,5-dioxo-1,3-dioxa-2-lanthanacyclopentan-2-yl oxalate decahydrate
PubChem CID 91886560
IUPAC Name lanthanum;oxalic acid;decahydrate
SMILES [La+3].[La+3].[O-]C(=O)C([O-])=O.[O-]C(=O)C([O-])=O.[O-]C(=O)C([O-])=O

Specifications

Melting Point (decomposition)
Physical Form Powder
Quantity 100 g
Linear Formula La2(C2O4)3·xH2O
UN Number UN3288
Solubility Information Soluble in water. Soluble in strong mineral acids.
Formula Weight 541.87 (Anhydrous)
Sensitivity Hygroscopic
Chemical Name or Material Lanthanum (III) oxalate hydrate
Hazard Category H302+H312
Hazard Statement GHS H Statement
H302-H312
Harmful if swallowed.
Harmful in contact with skin.
Precautionary Statement P264b-P270-P280-P301+P312-P302+P352-P312-P330-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLID, INORGANIC, N.O.S.
EINECSNumber 208-656-1
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.