Learn More
Strem, An Ascensus Company CAS# 52093-26-2. 10g. Lanthanum(III) trifluoromethanesulfonate, min. 97% (Lanthanum triflate). MFCD00077560
SDP
Catalog No. 5090112738
Encompass
CAS# 52093-26-2. 10g. Lanthanum(III) trifluoromethanesulfonate, min. 97% (Lanthanum triflate). MFCD00077560. Molecular Weight: 586.11. Molecular Formula: La(SO3CF3)3. Color/form: white pwdr. Strem# 57-2800. http://www.strem.com/catalog/v/57-2800/
Catalog No. 50-901-12738 Supplier Strem, An Ascensus Company Supplier No. 57280010G
CAS# 52093-26-2. 10g. Lanthanum(III) trifluoromethanesulfonate, min. 97% (Lanthanum triflate). MFCD00077560. Molecular Weight: 586.11. Molecular Formula: La(SO3CF3)3. Color/form: white pwdr. Strem# 57-2800. http://www.strem.com/catalog/v/57-2800/