Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lanthanum nickel powder, -100 mesh, 99.5% (metals basis)

Catalog No. AA8913306
Click to view available options
Quantity:
5 g
25 g

Catalog No. AA8913306

Supplier: Thermo Scientific Chemicals 08913306

Only null left
Add to Cart

CAS: 12196-72-4 | LaNi5 | 432.37 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under Argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12196-72-4
Molecular Formula LaNi5
Molecular Weight (g/mol) 432.37
MDL Number MFCD00198177
InChI Key BUCVQDCWULHLJZ-UHFFFAOYSA-N
Synonym lanthanum nickel, lani5, lanthanum pentanickel, lanthanum, compd. with nickel 1:1, lanthanum-nickel alloy, acmc-1brhe, lanthanum nickel, reacton, lanthanum nickel powder,-100 mesh, lanthanum nickel eutectic pieces, reacton, hy-stor™ 205 lanthanum-nickel alloy
PubChem CID 14367487
IUPAC Name lanthanum;nickel
SMILES [Ni].[Ni].[Ni].[Ni].[Ni].[La]

Specifications

Quantity 5 g
Mesh Size −100 Mesh
Odor Odorless
Sensitivity Air and moisture sensitive
UN Number UN3178
Percent Purity 99.5%
Assay (metals basis)
Chemical Name or Material Lanthanum nickel powder
Hazard Category H228-H334-H350-H372
Hazard Statement GHS H Statement
H228-H260-H350-H317
Flammable solid.
In contact with water releases flammable gases which may ignite spontaneously.
May cause cancer.
May cause an allergic skin reaction.
Precautionary Statement P201-P202-P210-P240-P241-P260-P264b-P270-P281-P285-P304+P341-P308+P313-P342+P311-P370+P378q-P501c
DOTInformation Hazard Class: 4.1; Packaging Group: III
EINECSNumber 235-372-5
TSCA Yes
storageNote1
  • Packaged under Argon
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.