Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lanthanum Standard, 1000μg/mL in water with dilute nitric acid, Ultra Scientific

Catalog No. USICP057
Encompass
Change view
Click to view available options
Quantity:
125 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
USICP057 125 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. USICP057 Supplier Agilent Technologies Supplier No. ICP057
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7439-91-0 , 7697-37-2 , 7732-18-5
Molecular Formula La
Molecular Weight (g/mol) 138.91
MDL Number MFCD00011066
InChI Key FZLIPJUXYLNCLC-UHFFFAOYSA-N
PubChem CID 23926
ChEBI CHEBI:33336
IUPAC Name lanthanum
SMILES [La]

Specifications

CAS Max % 98.9, 1.0, 0.1
Analyte or Component Names Lanthanum,water,nitric acid
Element Nitric Acid, La
Solvent or Matrix Water/Dilute Nitric Acid
Quantity 125 mL
Concentration or Composition (by Analyte or Components) 1000μg/mL
Chemical Name or Material Lanthanum Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.