Learn More
Sigma Aldrich Lanthanum strontium cobalt ferrite
Catalog No. 70428810G Shop All MilliporeSigma Sigma Organic Chemistry Products
Click to view available options
Quantity:
10 g
Catalog No. Quantity
70-428-810G 10 g
1 options
Catalog No. 70-428-810G Supplier Sigma Aldrich Supplier No. 70428810G
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Lanthanum strontium cobalt ferrite (LSCF 6428) is a promising cathode material, which has a low operating temperature. It has a high ionic and electronic conductivity with a high electrocatalytic activity.LSCF-6428 may be used as the cathode in solid oxide fuel cells (SOFCs).
Specifications
Quantity 10 g
Synonym LCSF; Lanthanum strontium cobalt iron oxide
Biological Activity Respiratory system