Learn More
Sigma Aldrich Lanthanum strontium cobalt ferrite
Catalog No. 73463210G Shop All MilliporeSigma Sigma Organic Chemistry Products
Click to view available options
Quantity:
10 g
Catalog No. Quantity
73-463-210G 10 g
1 options
Catalog No. 73-463-210G Supplier Sigma Aldrich Supplier No. 73463210G
Add to Cart
Add to Cart
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Lanthanum strontium cobalt ferrite (LSCF) is a mixed ionic and electron conducting pervoskite. The ionic conductivity is lower than YSZ although maintaining high conductivity. X-ray diffraction and electrochemical measurements of an operational LSCF cathode was conducted to study its lattice expansion during use. LSCF can be prepared by solid state reaction of individual cation oxides.LSCF oxygen electrodes can be used in the fabrication of high-performance solid oxide fuel cells (SOFCs).
Specifications
Quantity 10 g
Biological Activity Respiratory system