Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Lanthanum strontium cobaltite, LSC-82

Catalog No. 76369125G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
76-369-125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 76-369-125G Supplier Sigma Aldrich Supplier No. 76369125G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

LSC-82 is a promising cathode material for Solid Oxide Fuel Cells with a pervoskite type structure.LSC-82 has the unique ability to reduce itself reversibly at moderate partial pressures of oxygen, thus producing large numbers of oxygen ion vacancies. The ion vacancies allow a number of events to take place, such as; ease the transport of oxygen ion and cause oxygen surface exchange, raise mechanical stress by causing volume expansion of the lattice.Influence of change in temperature on the properties of LSC-82 has been reported in detail. Effect of temperature on magnetic susceptibility has also been reported.SOFCs containing LSC-82 cathodes show good performances with doped ceria electrolytes compared to yttria stabilized zirconia electrolytes.

Specifications

CAS 108916-09-2
Molecular Formula La0.8 Sr0.2 CoO3
Linear Formula La0.8 Sr0.2 CoO3
Quantity 25 g
Synonym Cobalt lanthanum strontium oxide; LSC; LSC-82; LSCo; Lanthanum strontium cobalt oxide; Strontium doped lanthanum cobalt oxide
Molecular Weight (g/mol) 235.58
Percent Purity 99% trace metals basis
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.